Gallium metal ≥99.999% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
44418-22
44418-36
CAAA44418-22EA
783.68
CAD
CAAA44418-22
CAAA44418-36
Gallium metal ≥99.999% (metals basis)
Gallium
Purity: 99.999%
CAS No.: 7440-55-3
Physical Form: May be solid or liquid due to ambient temperature
UN No.: 2803
MDL No.: MFCD00134045
Formula:
Ga Boiling Pt: 2403 °C (1013 hPa) Melting Pt: 29 °C Density: 5.904 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134045 CAS Number: 7440-55-3 EINECS: 231-163-8 UN: 2803 ADR: 8,III Merck Index: 12,04364 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...