Oxalic acid, anhydrous ≥98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Ethanedioic acid
44410-18
44410-30
CAAA44410-18EA
62.57
CAD
CAAA44410-18
CAAA44410-30
Oxalic acid, anhydrous ≥98%
Oxalic acid
Purity: 98%
CAS No.: 144-62-7
Molecular Formula: C2H2O4
Formula Weight: 90.04
Physical Form: Crystalline
Density: 1.9
Appearance: White
pH: 1.3
Melting Point: ca 185° dec.
Beilstein Reference: 385686
Merck Reference: 14,6911
UN No.: 3261
MDL No.: MFCD00002573
Formula:
C₂H₂O₄ Boiling Pt: 365 °C (1013 hPa) Melting Pt: ∼ 185 °C (dec.) Density: 1.9 Storage Temperature: Ambient |
MDL Number:
MFCD00002573 CAS Number: 144-62-7 EINECS: 205-634-3 UN: 3261 ADR: 8,III Merck Index: 14,6911 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...