Silicon ≥98%, crystalline powder APS ≤50 nm, laser synthesised from vapor phase
Supplier: Thermo Scientific Chemicals
|
Warning
|
44384-09
44384-18
CAAA44384-09EA
470.84
CAD
CAAA44384-09
CAAA44384-18
Silicon ≥98%, crystalline powder APS ≤50 nm, laser synthesised from vapor phase
Silicon
Purity: 98%
CAS No.: 7440-21-3
Physical Form: S.A. 70-100m2/g
Appearance: White
Odor: Odorless
UN No.: 1346
MDL No.: MFCD00085311
Formula:
Si Storage Temperature: Ambient |
MDL Number:
MFCD00085311 CAS Number: 7440-21-3 EINECS: 231-130-8 UN: 1346 ADR: 4.1,III Merck Index: 12,08635 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...