Europium(III) oxide ≥99.996% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals

44347-06 44347-14 44347-22
CAAAA44347-06EA 308.1 CAD
CAAAA44347-06 CAAAA44347-14 CAAAA44347-22
Europium(III) oxide ≥99.996% (REO, rare earth oxide basis), REacton®
Europium (III) oxide

Purity: 99.996% CAS No.: 1308-96-9 Molecular Formula: Eu2O3 Formula Weight: 351.92 Physical Form: Powder Sensitivity: Hygroscopic Merck Reference: 14,3904 MDL No.: MFCD00010997

Formula: Eu₂O₃
MW: 351.92 g/mol
Melting Pt: 2291 °C
Density: 7.42 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010997
CAS Number: 1308-96-9
EINECS: 215-165-6
Merck Index: 14,3904

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR