Cobalt(II) acetate tetrahydrate 98.0-102.0% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
44344-14
44344-22
44344-36
CAAAA44344-14EA
60.51
CAD
CAAAA44344-14
CAAAA44344-22
CAAAA44344-36
Cobalt(II) acetate tetrahydrate 98.0-102.0% ACS
Cobalt (II) acetate tetrahydrate
Purity: 98.0-102.0%
CAS No.: 6147-53-1
Molecular Formula: C4H6CoO4.4H2O
Formula Weight: 249.08 (177.02anhy)
Physical Form: Crystalline
Density: 1.71
Refractive Index: 1.542
Sensitivity: Hygroscopic
Appearance: Red
Odor: Odorless
Melting Point: ca 140° -4H2O
Beilstein Reference: 3731397
Merck Reference: 14,2433
UN No.: 3077
MDL No.: MFCD00149569
Formula:
(H₃CCOO)₂Co·4H₂O Melting Pt: ∼140 °C (-4H₂O) Density: 1.71 Storage Temperature: Ambient |
MDL Number:
MFCD00149569 CAS Number: 6147-53-1 EINECS: 200-755-8 UN: 3077 ADR: 9,III Merck Index: 14,2433 |
Specification Test Results
Insoluble matter | 0.01% |
Cl | 0.002% |
Cu | 0.002% |
Fe | 0.001% |
PB | 0.001% |
MG | 0.005% |
Ni | 0.001 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...