Lithium perchlorate, anhydrous 99%

Supplier: Thermo Scientific Chemicals
Danger

44225-09 44225-18 44225-30
CAAA44225-09EA 83.42 CAD
CAAA44225-09 CAAA44225-18 CAAA44225-30
Lithium perchlorate, anhydrous 99%
Lithium perchlorate

Lithium battery


Purity: 99% CAS No.: 7791-03-9 Molecular Formula: ClLiO4 Formula Weight: 106.40 Physical Form: Granular, Ampouled under argon Density: 2.43 Appearance: White Melting Point: 236° Boiling Point: 400° dec. Merck Reference: 14,5539 UN No.: 1481 MDL No.: MFCD00011079


Lithium battery

Formula: ClLiO₄
MW: 106.39 g/mol
Boiling Pt: 400 °C (dec.)
Melting Pt: 236 °C
Density: 2.43
Storage Temperature: Ambient
MDL Number: MFCD00011079
CAS Number: 7791-03-9
EINECS: 232-237-2
UN: 1481
ADR: 5.1,II
Merck Index: 14,5539

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR