Rubidium ≥99.75% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

44214-03 44214-06
CAAA44214-03EA 161.16 CAD
CAAA44214-03 CAAA44214-06
Rubidium ≥99.75% (metals basis)
Rubidium

Purity: 99.75% CAS No.: 7440-17-7 Physical Form: Ampouled under argon UN No.: 1423 MDL No.: MFCD00134055

Formula: Rb
MW: 85.47 g/mol
Boiling Pt: 700 °C (1013 hPa)
Melting Pt: 39 °C
Density: 0.83 g/cm³ (1399.85 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134055
CAS Number: 7440-17-7
EINECS: 231-126-6
UN: 1423
ADR: 4.3,I
Merck Index: 12,08439

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR