Tungsten ≥99.95% (metals basis), powder APS <1 µm
Supplier: Thermo Scientific Chemicals
|
Danger
|
44210-22
44210-36
44210-A3
CAAA44210-22EA
192.76
CAD
CAAA44210-22
CAAA44210-36
CAAA44210-A3
Tungsten ≥99.95% (metals basis), powder APS <1 µm
Tungsten
Purity: 99.95%
CAS No.: 7440-33-7
Appearance: Dark grey
UN No.: 3089
MDL No.: MFCD00011461
Formula:
W MW: 183.84 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011461 CAS Number: 7440-33-7 EINECS: 231-143-9 UN: 3089 ADR: 4.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...