Titanium ≥99.6% (metals basis) Grade 1, foil, annealed, Thickness: 0.05 mm (0.002 in)

Supplier: Thermo Scientific Chemicals
Warning

44202-FI 44202-GH 44202-GT
CAAA44202-FIEA 169.06 CAD
CAAA44202-FI CAAA44202-GH CAAA44202-GT
Titanium ≥99.6% (metals basis) Grade 1, foil, annealed, Thickness: 0.05 mm (0.002 in)
Titanium

Purity: 99.6% CAS No.: 7440-32-6 Physical Form: ≈0.14g/25x25mm, Temper:soft Odor: Odorless MDL No.: MFCD00011264

Formula: Ti
MW: 47.88 g/mol
Boiling Pt: 3262
Melting Pt: 1660
Density: 4.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR