Zinc chloride, ultra dry ≥99.999% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Zinc dichloride, Dichlorozinc, Zinc (II) chloride
44198-06
44198-14
44198-22
CAAA44198-06EA
146.47
CAD
CAAA44198-06
CAAA44198-14
CAAA44198-22
Zinc chloride, ultra dry ≥99.999% (metals basis)
Zinc chloride
Purity: 99.999%
CAS No.: 7646-85-7
Molecular Formula: ZnCl2
Formula Weight: 136.29
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 2.91
Melting Point: 290°
Boiling Point: 732°
Merck Reference: 14,10132
UN No.: 2331
MDL No.: MFCD00011295
Formula:
ZnCl₂ MW: 136.3 g/mol Boiling Pt: 732 Melting Pt: 283 Density: 2.91 Storage Temperature: Ambient |
MDL Number:
MFCD00011295 CAS Number: 7646-85-7 EINECS: 231-592-0 UN: 2331 ADR: 8 III Merck Index: 14,10132 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...