Silicon ≥99.9% (metals basis), powder APS 1-5 µm
Supplier: Thermo Scientific Chemicals
|
Warning
|
44185-09
44185-18
44185-30
CAAA44185-09EA
144.41
CAD
CAAA44185-09
CAAA44185-18
CAAA44185-30
Silicon ≥99.9% (metals basis), powder APS 1-5 µm
Silicon
Purity: 99.9%
CAS No.: 7440-21-3
Physical Form: Powder
Appearance: White
Odor: Odorless
UN No.: 1346
MDL No.: MFCD00085311
Formula:
Si Storage Temperature: Ambient |
MDL Number:
MFCD00085311 CAS Number: 7440-21-3 EINECS: 231-130-8 UN: 1346 ADR: 4.1,III Merck Index: 12,08635 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...