Neodymium ≥99.8% (REO, rare earth oxide basis), powder -40 mesh

Supplier: Thermo Scientific Chemicals
Danger

44181-06 44181-14
CAAA44181-06EA 175.38 CAD
CAAA44181-06 CAAA44181-14
Neodymium ≥99.8% (REO, rare earth oxide basis), powder -40 mesh
Neodymium

Purity: 99.9% CAS No.: 7440-00-8 Physical Form: Ampouled under argon Sensitivity: Air and Moisture Sensitive Appearance: Grey to silver Odor: Odorless UN No.: 3089 MDL No.: MFCD00011130

Formula: Nd
Boiling Pt: 2474 °C (1013 hPa)
Melting Pt: 1014.85 °C
Density: 6.46 g/cm³ (1240 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011130
CAS Number: 7440-00-8
EINECS: 231-109-3
UN: 3089
ADR: 4.1,II
Merck Index: 12,06538

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR