Erbium ≥99.9% (REO, rare earth oxide basis), powder -40 mesh
Supplier: Thermo Scientific Chemicals
|
Danger
|
44169-06
44169-14
CAAA44169-06EA
360.24
CAD
CAAA44169-06
CAAA44169-14
Erbium ≥99.9% (REO, rare earth oxide basis), powder -40 mesh
Erbium
Purity: 99.9%
CAS No.: 7440-52-0
Physical Form: Packaged under argon
Sensitivity: Air and Moisture Sensitive
Appearance: Gray
UN No.: 3089
MDL No.: MFCD00010987
Formula:
Er Boiling Pt: 2863 °C (1013 hPa) Melting Pt: 1529 °C Density: 9.066 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010987 CAS Number: 7440-52-0 EINECS: 231-160-1 UN: 3089 ADR: 4.1,II Merck Index: 12,03679 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...