Yttrium ≥99.6% (REO, rare earth oxide basis), powder -40 mesh

Supplier: Thermo Scientific Chemicals
Danger

44147-06 44147-14
CAAA44147-06EA 189.6 CAD
CAAA44147-06 CAAA44147-14
Yttrium ≥99.6% (REO, rare earth oxide basis), powder -40 mesh
Yttrium

Purity: 99.6% CAS No.: 7440-65-5 Physical Form: Packaged under argon Odor: Odorless UN No.: 3089 MDL No.: MFCD00011468

Formula: Y
MW: 88.91
Boiling Pt: 2927
Melting Pt: 1525
Density: 4.472
Storage Temperature: Ambient
MDL Number: MFCD00011468
CAS Number: 7440-65-5
EINECS: 231-174-8
UN: 3089
ADR: 4.1 II
Merck Index: 12,10240

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR