Tetrabutylammonium perchlorate electrochemical grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
43999-09
43999-18
43999-30
CAAA43999-09EA
85.32
CAD
CAAA43999-09
CAAA43999-18
CAAA43999-30
Tetrabutylammonium perchlorate electrochemical grade
Tetrabutylammonium perchlorate
Suitable for use as a polarographic supporting electrolyte
CAS No.: 1923-70-2
Molecular Formula: C16H36ClNO4
Formula Weight: 341.92
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: White
Melting Point: 210-217°
Solubility: Soluble in ethanol. Very slightly soluble in water
UN No.: 1481
MDL No.: MFCD00038722
Suitable for use as a polarographic supporting electrolyte
Formula:
C₁₆H₃₆ClNO₄ Melting Pt: 210…217 °C Density: 1.07 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00038722 CAS Number: 1923-70-2 EINECS: 217-655-5 UN: 1481 ADR: 5.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...