Cerium ≥99% (REO, rare earth oxide basis), ingot

Supplier: Thermo Scientific Chemicals
Danger

43977-30 43977-A1
CAAA43977-30EA 238.58 CAD
CAAA43977-30 CAAA43977-A1
Cerium ≥99% (REO, rare earth oxide basis), ingot
Cerium

Purity: 99% CAS No.: 7440-45-1 Physical Form: Packaged under mineral oil Appearance: Gray Odor: Odorless UN No.: 1333 MDL No.: MFCD00010924

Formula: Ce
MW: 140.12 g/mol
Boiling Pt: 3468 °C (1013 hPa)
Melting Pt: 795 °C
Density: ∼6.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010924
CAS Number: 7440-45-1
EINECS: 231-154-9
UN: 1333
ADR: 4.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR