Tellurium ≥99.999% (metals basis), powder, Puratronic® -18/+60 mesh

Supplier: Thermo Scientific Chemicals
Danger

43960-18 43960-30
CAAA43960-18EA 289.14 CAD
CAAA43960-18 CAAA43960-30
Tellurium ≥99.999% (metals basis), powder, Puratronic® -18/+60 mesh
Tellurium

Purity: 99.999% CAS No.: 13494-80-9 Odor: Odorless UN No.: 3288 MDL No.: MFCD00134062

Formula: Te
Boiling Pt: 990 °C (1013 hPa)
Melting Pt: 450 °C
Density: 8.83 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134062
CAS Number: 13494-80-9
EINECS: 236-813-4
UN: 3288
ADR: 6.1,III
Merck Index: 12,09273

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR