Dipotassium hexabromopalladate ≥99.999% (metals basis) Pd ≥ 15.5%, Premion®
Supplier: Thermo Scientific Chemicals
|
Warning
|
43947-03
43947-06
CAAAA43947-03EA
180.12
CAD
CAAAA43947-03
CAAAA43947-06
Dipotassium hexabromopalladate ≥99.999% (metals basis) Pd ≥ 15.5%, Premion®
Dipotassium hexabromopalladate
Purity: 99.999%
CAS No.: 16919-74-7
Molecular Formula: K2PdBr6
Formula Weight: 664.04
Physical Form: Crystalline
Formula:
Br₆K₂Pd Storage Temperature: Ambient |
MDL Number:
MFCD03093651 CAS Number: 16919-74-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...