Potassium disulfite ≥95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
43908-22
43908-36
43908-A3
CAAAA43908-22EA
60.99
CAD
CAAAA43908-22
CAAAA43908-36
CAAAA43908-A3
Potassium disulfite ≥95%
di-Potassium disulphite
Purity: 95+%
CAS No.: 16731-55-8
Molecular Formula: K2S2O5
Formula Weight: 222.32
Physical Form: Powder
Sensitivity: Air Sensitive
Appearance: White
Odor: Pungent
Melting Point: 190° dec.
Merck Reference: 14,7645
MDL No.: MFCD00167605
Formula:
K₂O₅S₂ Melting Pt: 190 °C (dec.) Density: 1.2 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00167605 CAS Number: 16731-55-8 EINECS: 240-795-3 Merck Index: 14,7645 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...