1-Propanol ≥99.5% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
43848-AP
43848-K2
43848-K7
43848-M6
CAAA43848-APEA
74.58
CAD
CAAA43848-AP
CAAA43848-K2
CAAA43848-K7
CAAA43848-M6
1-Propanol ≥99.5% ACS
1-Propanol
Purity: 99.5+%
CAS No.: 71-23-8
Molecular Formula: CH3CH2CH2OH
Formula Weight: 60.10
Flash Point: 15°(59°F)
Physical Form: Liquid
Density: 0.804
Refractive Index: 1.3850
Odor: Alcohol-like
Melting Point: -127°
Boiling Point: 97°
Beilstein Reference: 1098242
Merck Reference: 14,7842
UN No.: 1274
MDL No.: MFCD00002941
Formula:
H₃CCH₂CH₂OH Boiling Pt: 97 °C Melting Pt: –127 °C Density: 0.804 Flash Pt: 15 °C (59 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002941 CAS Number: 71-23-8 EINECS: 200-746-9 UN: 1274 ADR: 3,II Merck Index: 14,7842 |
|
Specification Test Results
Color (APHA) | 10 |
Residue after evaporation | 0.001% |
Titratable acid | 0.0004meq/g |
Isopropyl alcohol | 0.05% |
H2O | 0.2% |
Ethanol (CH3CH2OH) | 0.01% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...