5-Fluorosalicylaldehyde 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
43844-03
43844-06
CAAA43844-03EA
156.1
CAD
CAAA43844-03
CAAA43844-06
5-Fluorosalicylaldehyde 98+%
5-Fluorosalicylaldehyde
Purity: 98+%
CAS No.: 347-54-6
Molecular Formula: C7H5FO2
Formula Weight: 140.12
Physical Form: Powder/flakes
Sensitivity: Air Sensitive
MDL No.: MFCD01090997
Formula:
C₇H₅FO₂ Melting Pt: 82…85 °C Storage Temperature: Ambient |
MDL Number:
MFCD01090997 CAS Number: 347-54-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...