Aluminium oxide, γ-phase, high surface area catalyst support bimodal

Supplier: Thermo Scientific Chemicals

43832-18 43832-30 43832-A1 43832-A7
CAAAA43832-18EA 47.56 CAD
CAAAA43832-18 CAAAA43832-30 CAAAA43832-A1 CAAAA43832-A7
Aluminium oxide, γ-phase, high surface area catalyst support bimodal
Aluminium oxide, γ-phase

S.A. 255 m2/g, median pore 70 micron and 5000A, total pore volume 1.14cc/g, packing density 0.395 kg/L


Physical Form: 1/8" pellets
Odour: Odourless


S.A. 255m2/g, median pore 70 micron and 5000A, total pore volume 1.14cc/g, packing density 0.395kg/L

Formula: Al₂O₃
MW: 101.96 g/mol
Boiling Pt: 2980
Melting Pt: 2045
Density: 3.965
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
Merck Index: 14,356

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR