Titanium(IV) oxide anatase catalyst support
Supplier: Thermo Scientific Chemicals
|
Warning
|
43828-30
43828-A1
43828-A7
CAAAA43828-30EA
120.08
CAD
CAAAA43828-30
CAAAA43828-A1
CAAAA43828-A7
Titanium(IV) oxide anatase catalyst support
Titanium (IV) oxide anatase
CAS No.: 1317-80-2
Molecular Formula: TiO2
Formula Weight: 79.90
Physical Form: 1/8" Pellets
Appearance: White
Odor: Odorless
MDL No.: MFCD00011269
Formula:
TiO₂ Boiling Pt: 2900 °C (1013 hPa) Melting Pt: 1855 °C Density: 4.2 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011269 CAS Number: 1317-70-0 EINECS: 215-280-1 Merck Index: 13,09549 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...