1,6-Diisocyanatohexane 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
43803-18
43803-30
CAAA43803-18EA
67.62
CAD
CAAA43803-18
CAAA43803-30
1,6-Diisocyanatohexane 98+%
1,6-Diisocyanatohexane
Purity: 98+%
CAS No.: 822-06-0
Molecular Formula: OCN(CH2)6NCO
Formula Weight: 168.19
Flash Point: 130°(266°F)
Physical Form: Liquid, Packaged under argon
Density: 1.05
Refractive Index: 1.4520
Sensitivity: Moisture Sensitive
Odor: Pungent
Melting Point: -67°
Boiling Point: 255°
Beilstein Reference: 956709
UN No.: 2281
MDL No.: MFCD00002047
Formula:
C₈H₁₂N₂O₂ Boiling Pt: 255 °C Melting Pt: –67 °C Density: 1.05 Flash Pt: 130 °C (266 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002047 CAS Number: 822-06-0 EINECS: 212-485-8 UN: 2281 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...