Pyridine, anhydrous ≥99.5%, ChemSeal™
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Azabenzene, Azine
Packaged under argon in resealable ChemSeal™ bottles.
43799-AE
43799-K2
CAAA43799-AEEA
131.14
CAD
CAAA43799-AE
CAAA43799-K2
Pyridine, anhydrous ≥99.5%, ChemSeal™
Pyridine
Purity: 99.5+%
CAS No.: 110-86-1
Molecular Formula: C5H5N
Formula Weight: 79.10
Flash Point: 20°(68°F)
Physical Form: Liquid
Density: 0.978
Refractive Index: 1.5100
Odor: Unpleasant
Melting Point: -42°
Boiling Point: 115°
Beilstein Reference: 103233
Merck Reference: 14,7970
UN No.: 1282
MDL No.: MFCD00011732
Formula:
C₅H₅N MW: 79.1 g/mol Boiling Pt: 115 °C Melting Pt: –42 °C Density: 0.978 Flash Pt: 20 °C (68 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00011732 CAS Number: 110-86-1 EINECS: 203-809-9 UN: 1282 ADR: 3,II Merck Index: 14,7970 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...