Propagyl amine hydrochloride ≥95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
43775-03
43775-06
CAAA43775-03EA
65.25
CAD
CAAA43775-03
CAAA43775-06
Propagyl amine hydrochloride ≥95%
Propagyl amine hydrochloride
Purity: 95%
CAS No.: 15430-52-1
Molecular Formula: HC≡CCH2NH2.HCl
Formula Weight: 91.54
Physical Form: Powder
Sensitivity: Hygroscopic
UN No.: 2811
MDL No.: MFCD00012907
Formula:
C₃H₆ClN Melting Pt: 179…182 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012907 CAS Number: 15430-52-1 EINECS: 239-440-5 UN: 2811 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...