Boron nitride, paste

Supplier: Thermo Scientific Chemicals
Warning

43773-14 43773-22 43773-36
CAAAA43773-14EA 92.11 CAD
CAAAA43773-14 CAAAA43773-22 CAAAA43773-36 CAAAA43773-A3
Boron nitride, paste
Boron nitride

Resistance against molten metals and salts, high thermal conductivity and dielectric strength


Physical Form: Paste Density: 1.40 MDL No.: MFCD00011317


Resistance against molten metals and salts, high thermal conductivity and dielectric strength

Formula: BN
MW: 24.82 g/mol
Melting Pt: 2527
Density: 1.40
Storage Temperature: Ambient
MDL Number: MFCD00011317
CAS Number: 10043-11-5
EINECS: 233-136-6
Merck Index: 13,01336

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR