Sodium hydroxide 25% (w/v) in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

43771-K2 43771-K7
CAAAA43771-K2EA 62.73 CAD
CAAAA43771-K2 CAAAA43771-K7
Sodium hydroxide 25% (w/v) in aqueous solution
Sodium hydroxide

CAS No.: 1310-73-2 Molecular Formula: NaOH Formula Weight: 40.00 Physical Form: Liquid UN No.: 1824 MDL No.: MFCD00003548

Formula: NaOH
Boiling Pt: 119…135 °C (1013 hPa)
Melting Pt: 9…10 °C
Density: 1.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR