1,2-Bis(dimethylsilyl)benzene 98%
Supplier: Thermo Scientific Chemicals
Warning
|
Synonyms:
1,2-Phenylenebis(dimethylsilane)
43755-04
43755-09
CAAAA43755-02EA
78.29
CAD
CAAAA43755-02
CAAAA43755-04
CAAAA43755-09
1,2-Bis(dimethylsilyl)benzene 98%
1,2-Bis(dimethylsilyl)benzene
Purity: 98%
CAS No.: 17985-72-7
Molecular Formula: C10H18Si2
Formula Weight: 194.43
Flash Point: 70°(158°F)
Physical Form: Liquid
Density: 0.898
Refractive Index: 1.5140
Boiling Point: 101°/13mm
MDL No.: MFCD00142462
Formula:
C₁₀H₁₈Si₂ MW: 194.42 g/mol Boiling Pt: 101 °C (13 mmHg) Density: 0,898 Flash Pt: 70 °C (158 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00142462 CAS Number: 17985-72-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...