Tantalum pentachloride, anhydrous ≥99.995% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Danger

43731-06 43731-14 43731-22
CAAA43731-06EA 155.47 CAD
CAAA43731-06 CAAA43731-14 CAAA43731-22
Tantalum pentachloride, anhydrous ≥99.995% (metals basis), Puratronic®
Tantalum pentachloride

Purity: 99.995% CAS No.: 7721-01-9 Molecular Formula: TaCl5 Formula Weight: 358.21 Physical Form: Powder, Ampouled under argon Density: 3.68 Appearance: White to pale yellow Odor: Acidic Melting Point: 216° Boiling Point: 242° Merck Reference: 14,9054 UN No.: 3260 MDL No.: MFCD00011253

Formula: Cl₅Ta
Boiling Pt: 242 °C
Melting Pt: 216 °C
Density: 3.68
Storage Temperature: Ambient
MDL Number: MFCD00011253
CAS Number: 7721-01-9
EINECS: 231-755-6
UN: 3260
ADR: 8,III
Merck Index: 14,9054

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR