Niobium(V) chloride ≥99.999% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Niobium pentachloride
43729-06
43729-14
43729-22
CAAA43729-06EA
181.7
CAD
CAAA43729-06
CAAA43729-14
CAAA43729-22
Niobium(V) chloride ≥99.999% (metals basis), Puratronic®
Niobium (V) chloride
Purity: 99.999%
CAS No.: 10026-12-7
Molecular Formula: NbCl5
Formula Weight: 270.20
Physical Form: Powder, Packaged under argon
Density: 2.74
Melting Point: 208.3°
Boiling Point: 248.2°
Merck Reference: 14,6557
UN No.: 3260
MDL No.: MFCD00011127
Formula:
Cl₅Nb Boiling Pt: 248.2 °C Melting Pt: 208.3 °C Density: 2.74 Storage Temperature: Ambient |
MDL Number:
MFCD00011127 CAS Number: 10026-12-7 EINECS: 233-059-8 UN: 3260 ADR: 8,III Merck Index: 14,6557 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...