3-Hydroxy-3-methylbutanone 90+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
43724-06
43724-14
43724-22
CAAA43724-06EA
70.63
CAD
CAAA43724-06
CAAA43724-14
CAAA43724-22
3-Hydroxy-3-methylbutanone 90+%
3-Hydroxy-3-methylbutanone
Purity: 90+%
CAS No.: 115-22-0
Molecular Formula: C5H10O2
Formula Weight: 102.13
Flash Point: 42°(107.6°F)
Physical Form: Liquid
Density: 0.96
Refractive Index: 1.4150
Boiling Point: 140-141°
UN No.: 1224
MDL No.: MFCD00004460
Formula:
C₅H₁₀O₂ Boiling Pt: 140…141 °C Density: 0.96 Flash Pt: 42 °C (107.6 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004460 CAS Number: 115-22-0 EINECS: 204-073-1 UN: 1224 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...