Palladium ≥99.95% (metals basis), powder APS 0,5-1,7 micron

Supplier: Thermo Scientific Chemicals
Danger

43715-03 43715-06 43715-14
CAAA43715-03EA 522.98 CAD
CAAA43715-03 CAAA43715-06 CAAA43715-14
Palladium ≥99.95% (metals basis), powder APS 0,5-1,7 micron
Palladium

Purity: 99.95% CAS No.: 7440-05-3 Physical Form: Tap density 2.4-5.0g/cm3, S.A. 0.7-2.0m2/g Odor: Odorless UN No.: 3089 MDL No.: MFCD00011167

Formula: Pd
Density: 12.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6
UN: 3089
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR