2,3-Dimethylpentane 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
43707-06
43707-14
CAAA43707-06EA
97.96
CAD
CAAA43707-06
CAAA43707-14
2,3-Dimethylpentane 97%
2,3-Dimethylpentane
Purity: 97%
CAS No.: 565-59-3
Molecular Formula: C7H16
Formula Weight: 100.21
Flash Point: -6°(21°F)
Physical Form: Liquid
Density: 0.690
Refractive Index: 1.3920
Boiling Point: 89-90°
UN No.: 1206
MDL No.: MFCD00009337
Formula:
C₇H₁₆ Boiling Pt: 89…90 °C Melting Pt: –135 °C Density: 0.690 Flash Pt: –6 °C (21 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009337 CAS Number: 565-59-3 EINECS: 209-280-0 UN: 1206 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...