Nickel ≥99.8% (metals basis), spherical powder -400 mesh
Supplier: Thermo Scientific Chemicals
|
Danger
|
43704-14
43704-22
43704-36
43704-A3
CAAA43704-14EA
34.44
CAD
CAAA43704-14
CAAA43704-22
CAAA43704-36
CAAA43704-A3
Nickel ≥99.8% (metals basis), spherical powder -400 mesh
Nickel
Purity: 99.8+%
CAS No.: 7440-02-0
Odor: Odorless
UN No.: 3089
MDL No.: MFCD00011137
Formula:
Ni Boiling Pt: 2371 °C (1013 hPa) Melting Pt: 1455 °C Density: 8.9 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011137 CAS Number: 7440-02-0 EINECS: 231-111-4 UN: 3089 ADR: 4.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...