Palladium (II) sulfate dihydrate ≥99.995% (metals basis), Premion®
Supplier: Thermo Scientific Chemicals
|
Danger
|
43698-03
43698-06
CAAA43698-03EA
271.76
CAD
CAAA43698-03
CAAA43698-06
Palladium (II) sulfate dihydrate ≥99.995% (metals basis), Premion®
Palladium (II) sulphate dihydrate
Purity: 99.995%
CAS No.: 13444-98-9
Molecular Formula: O4PdS.2H2O
Formula Weight: 238.51 (202.48anhy)
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: Red to brown
Melting Point: dec.
UN No.: 3260
MDL No.: MFCD00011173
Formula:
O₄PdS·2H₂O MW: 238.51 Melting Pt: Dec. Storage Temperature: Ambient |
MDL Number:
MFCD00011173 CAS Number: 13444-98-9 EINECS: 236-957-8 UN: 3260 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...