Chloroform ≥99.5% stabilised for HPLC

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane

Stabilization: Stabilised with amylene

43685-K2 43685-K7 43685-M1 43685-M6
CAAA43685-K2EA 109.97 CAD
CAAA43685-K2 CAAA43685-K7 CAAA43685-M1 CAAA43685-M6
Chloroform ≥99.5% stabilised for HPLC
Chloroform

Purity: 99.5+% min CAS No.: 67-66-3 Molecular Formula: CHCl3 Formula Weight: 119.38 Physical Form: Liquid Density: 1.492 Refractive Index: 1.4460 Odor: Characteristic Melting Point: -63° Boiling Point: 60.5-61.5° Beilstein Reference: 1731042 Merck Reference: 14,2141 UN No.: 1888 MDL No.: MFCD00000826

Formula: CHCl₃
Boiling Pt: 60.5…61.5 °C
Melting Pt: –63 °C
Density: 1.492
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1,III
REACH: 05-2114614906-45
Merck Index: 14,2141
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR