Titanium ≥99.99% (metals basis), foil, stress relieved, Thickness: 0.5 mm (0.02 in)

Supplier: Thermo Scientific Chemicals

043676-GH 43676-FI 43676-FF 43676-GH 043676-FF 043676-FI
CAAA43676-GHEA 1568.94 CAD
CAAA43676-GH AA43676-FF CAAA43676-FF AA43676-FI AA43676-GH CAAA43676-FI
Titanium ≥99.99% (metals basis), foil, stress relieved, Thickness: 0.5 mm (0.02 in)
Titanium
Formula: Ti
MW: 47.88 g/mol
Boiling Pt: 3262 °C (1013 hPa)
Melting Pt: 1660 °C
Density: 4.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
Merck Index: 12,09610

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR