Gadolinium tribromide, ultra dry ≥99.99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Gadolinium (III) bromide
43673-06
43673-14
CAAAA43673-06EA
262.28
CAD
CAAAA43673-06
CAAAA43673-14
CAAAA43673-22
Gadolinium tribromide, ultra dry ≥99.99% (metals basis)
Gadolinium tribromide
Purity: 99.99%
CAS No.: 13818-75-2
Molecular Formula: GdBr3
Formula Weight: 396.96
Physical Form: Powder, Ampouled under argon
Sensitivity: Hygroscopic
MDL No.: MFCD00049741
Formula:
Br₃Gd MW: 396.96 g/mol Melting Pt: 770 °C Storage Temperature: Ambient |
MDL Number:
MFCD00049741 CAS Number: 13818-75-2 EINECS: 237-494-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...