Styrene-[D₈] (98% D) (Isotopic) stabilized
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Phenylethene-D8, Styrene-[²H₈], Vinylbenzene-D8, Phenylethylene-D8
Stabilization:
Stabilised with 4-tert-butylpyrocatechol (TBC)
Purity: 98%
CAS No.: 19361-62-7
Molecular Formula: C8D8
Formula Weight: 112.20
Storage Temperature: 2°C to 8°C
Flash Point: 31°(88°F)
Physical Form: Liquid, Ampouled under argon
Density: 0.979
Refractive Index: 1.5445
Sensitivity: Air Sensitive
Appearance: Colorless
Boiling Point: 50-51°/25mm
Beilstein Reference: 2327045
UN No.: 2055
MDL No.: MFCD00044231
Formula:
C₆D₅CDCD₂ MW: 112.09 g/mol Boiling Pt: 50…51 °C (25 mmHg) Melting Pt: –31 °C Density: 0.979 Flash Pt: 31 °C (88 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00044231 CAS Number: 19361-62-7 EINECS: 242-995-6 UN: 2055 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...