Tetrabutylammonium hydroxide 1.0 M in aqueous solution for HPLC

Supplier: Thermo Scientific Chemicals
Danger

43662-AC 43662-AE 43662-AP
CAAA43662-ACEA 92.75 CAD
CAAA43662-AC CAAA43662-AE CAAA43662-AP
Tetrabutylammonium hydroxide 1.0 M in aqueous solution for HPLC
Tetrabutylammonium hydroxide

Phase transfer catalyst, electrolyte in polarography, ion-pair extraction


CAS No.: 2052-49-5 Molecular Formula: C16H37NO Formula Weight: 259.48 Flash Point: None Physical Form: Liquid Sensitivity: Air Sensitive Beilstein Reference: 3571228 UN No.: 3267 MDL No.: MFCD00009425


Phase transfer catalyst, electrolyte in polarography, ion-pair extraction

Formula: C₁₆H₃₇NO
MW: 259.48
Melting Pt: 29.85
Density: 0.968
Storage Temperature: Ambient
MDL Number: MFCD00009425
CAS Number: 2052-49-5
EINECS: 218-147-6
UN: 3267
ADR: 8 II

Order Now

Specification Test Results

Color (APHA) 30
CO3 0.1%
Bu3N 0.2%
Cl 0.08%
K 2 ppm
NA 2 ppm
Polarographic test Pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR