Tin, powder_spherical APS ∼ 0.1 µm

Supplier: Thermo Scientific Chemicals
Warning

43461-14 43461-22
CAAA43461-14EA 429.76 CAD
CAAA43461-14 CAAA43461-22
Tin, powder_spherical APS ∼ 0.1 µm
Tin

CAS No.: 7440-31-5 Odor: Odorless UN No.: 3089 MDL No.: MFCD00133862

Formula: Sn
MW: 118.71 g/mol
Boiling Pt: 2270
Melting Pt: 232
Density: 7.31 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8
UN: 3089
ADR: 4.1 III
Merck Index: 12,09587

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR