Cadmium(II) iodide ≥99.999% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Danger
|
43442-14
43442-22
CAAA43442-14EA
169.06
CAD
CAAA43442-14
CAAA43442-22
Cadmium(II) iodide ≥99.999% (metals basis), Puratronic®
Cadmium (II) iodide
Purity: 99.999%
CAS No.: 7790-80-9
Molecular Formula: CdI2
Formula Weight: 366.21
Physical Form: Powder
Density: 5.67
Appearance: Pale brown
Odor: Odorless
Melting Point: 387-389°
Boiling Point: 787°
Merck Reference: 14,1621
UN No.: 2570
MDL No.: MFCD00010920
Formula:
CdI₂ Boiling Pt: 787 °C Melting Pt: 387…389 °C Density: 5.67 Storage Temperature: Ambient |
MDL Number:
MFCD00010920 CAS Number: 7790-80-9 EINECS: 232-223-6 UN: 2570 ADR: 6.1,III Merck Index: 14,1621 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...