Ytterbium ≥99.9% (REO, rare earth oxide basis), pieces, sublimated

Supplier: Thermo Scientific Chemicals

43431-06 43431-14 43431-22
CAAA43431-06EA 189.6 CAD
CAAA43431-06 CAAA43431-14 CAAA43431-22
Ytterbium ≥99.9% (REO, rare earth oxide basis), pieces, sublimated
Ytterbium

Purity: 99.9% CAS No.: 7440-64-4 Molecular Formula: Yb Formula Weight: 173.04 Physical Form: Packaged under argon

Formula: Yb
Boiling Pt: 1427 °C (1013 hPa)
Melting Pt: 824 °C
Density: 6.977 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011286
CAS Number: 7440-64-4
EINECS: 231-173-2
Merck Index: 12,10239

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR