2-Propanethiol 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Isopropyl mercaptan, Isopropanethiol, 2-Mercaptopropane
43422-AC
43422-AE
43422-AP
CAAA43422-ACEA
34.29
CAD
CAAA43422-AC
CAAA43422-AE
CAAA43422-AP
2-Propanethiol 98%
2-Propanethiol
Purity: 98%
CAS No.: 75-33-2
Molecular Formula: C3H8S
Formula Weight: 76.16
Flash Point: -34° (-29°F)
Physical Form: Liquid, Packaged under argon
Density: 0.82
Refractive Index: 1.4255
Sensitivity: Air Sensitive
Appearance: Colorless
Odor: Stench
Melting Point: -131°
Boiling Point: 57-60°
Beilstein Reference: 605260
UN No.: 2402
MDL No.: MFCD00004863
Formula:
C₃H₈S MW: 76.16 g/mol Boiling Pt: 57 Melting Pt: -130 Density: 0.82 Flash Pt: –34° (–29 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004863 CAS Number: 75-33-2 EINECS: 200-861-4 UN: 2402 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...