1-Butanethiol 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-Mercaptobutane, n-Butanethiol, NBM, n-Butyl mercaptan
43421-AC
43421-AE
CAAAA43421-ACEA
39.97
CAD
CAAAA43421-AC
CAAA43421-AE
1-Butanethiol 98%
1-Butanethiol
Purity: 98%
CAS No.: 109-79-5
Molecular Formula: C4H10S
Formula Weight: 90.19
Flash Point: 2°(36°F)
Physical Form: Liquid
Density: 0.842
Refractive Index: 1.4425
Sensitivity: Air Sensitive
Appearance: Colorless
Odor: Stench
Melting Point: -116°
Boiling Point: 98°
Beilstein Reference: 1730908
Merck Reference: 14,1577
UN No.: 2347
MDL No.: MFCD00004905
Formula:
C₄H₁₀S MW: 90.19 g/mol Boiling Pt: 98 Melting Pt: -116 Density: 0.842 Flash Pt: 2 °C (36 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004905 CAS Number: 109-79-5 EINECS: 203-705-3 UN: 2347 ADR: 3 II Merck Index: 14,1577 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...