Potassium chloride 3 M in aqueous solution

Supplier: Thermo Scientific Chemicals

Synonyms: Potassiumchloride

43398-AK 43398-K2
CAAAA43398-AKEA 65.43 CAD
CAAAA43398-AK CAAAA43398-K2
Potassium chloride 3 M in aqueous solution
Potassium chloride

For refilling calomel electrodes


CAS No.: 7447-40-7 Molecular Formula: KCl Formula Weight: 74.55 Physical Form: Liquid Odor: Odorless MDL No.: MFCD00011360


For refilling calomel electrodes

Formula: KCl
MW: 74.55 g/mol
Boiling Pt: 1420
Melting Pt: 773
Density: 1.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8
Merck Index: 14,07621

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR