Copper ≥99.9% (metals basis), spherical powder -170/+400 mesh oxygen 0.1%

Supplier: Thermo Scientific Chemicals
Warning

43385-22 43385-36 43385-A3
CAAA43385-22EA 85.27 CAD
CAAA43385-22 CAAA43385-36 CAAA43385-A3
Copper ≥99.9% (metals basis), spherical powder -170/+400 mesh oxygen 0.1%
Copper

Purity: 99.9% CAS No.: 7440-50-8 Appearance: Copper Odor: Odorless UN No.: 3089 MDL No.: MFCD00010965

Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
UN: 3089
ADR: 4.1 III
Merck Index: 13,02545

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR