1,3-Diethylbenzene 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
43369-AB
43369-S0
CAAA43369-ABEA
262.28
CAD
CAAA43369-AB
CAAA43369-S0
1,3-Diethylbenzene 97+%
1,3-Diethylbenzene
Purity: 97+%
CAS No.: 141-93-5
Molecular Formula: C10H14
Formula Weight: 134.22
Flash Point: 55°(131°F)
Physical Form: Liquid
Density: 0.86
Refractive Index: 1.4960
Appearance: Colorless
Odor: Aromatic
Boiling Point: 182°
UN No.: 2049
MDL No.: MFCD00009260
Formula:
C₁₀H₁₄ Boiling Pt: 182 °C Melting Pt: –84.2 °C Density: 0.86 Flash Pt: 55 °C (131 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009260 CAS Number: 141-93-5 EINECS: 205-511-4 UN: 2049 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...