Ruthenium(III) chloride hydrate ≥99.99% (metals basis), Premion®

Supplier: Thermo Scientific Chemicals
Danger

43364-04 43364-09
CAAA43364-04EA 279.66 CAD
CAAA43364-04 CAAA43364-09
Ruthenium(III) chloride hydrate ≥99.99% (metals basis), Premion®
Ruthenium (III) chloride hydrate

Purity: 99.99% CAS No.: 14898-67-0 Molecular Formula: RuCl3.xH2O Formula Weight: 207.43(anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: Black Melting Point: 100°dec Merck Reference: 14,8302 UN No.: 3260 MDL No.: MFCD00149844

Formula: Cl₃Ru·xH₂O
Melting Pt: 100 °C (dec.)
Density: 3.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149844
CAS Number: 14898-67-0
EINECS: 233-167-5
UN: 3260
ADR: 8,III
Merck Index: 14,8302

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR